Assay Details
| Gene Symbol: LOC100217256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143611.1 | NP_001137083.1 | 759 | 80 | ||
| GenBank mRNA | BT038218.1 | - | 759 | 80 | ||
More Information
Set Membership: |
|

LOC100217256
Genomic Map