Assay Details
| Gene Symbol: TLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111776.1 | NP_001105246.1 | 1137 | 101 | ||
| GenBank mRNA | AY496080.1 | - | 1137 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLK2| Gene Symbol: TLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111776.1 | NP_001105246.1 | 1137 | 101 | ||
| GenBank mRNA | AY496080.1 | - | 1137 | 101 | ||
Set Membership: |
|