Assay Details
| Gene Symbol: cycD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111579.1 | NP_001105049.1 | 993 | 72 | ||
| GenBank mRNA | AF351190.1 | - | 993 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cycD1| Gene Symbol: cycD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111579.1 | NP_001105049.1 | 993 | 72 | ||
| GenBank mRNA | AF351190.1 | - | 993 | 72 | ||
Set Membership: |
|