Assay Details
Gene Symbol: LOC100285065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157960.1 | NP_001151432.1 | 241 | 98 | ||
GenBank mRNA | EU970744.1 | - | 241 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
