Assay Details
| Gene Symbol: LOC100285035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157930.1 | NP_001151402.1 | 1045 | 109 | ||
| GenBank mRNA | EU970597.1 | - | 1045 | 109 | ||
More Information
Set Membership: |
|

LOC100285035
Genomic Map