Assay Details
| Gene Symbol: LOC100281316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154234.1 | NP_001147706.1 | 357 | 101 | ||
| GenBank mRNA | EU956307.1 | - | 357 | 101 | ||
More Information
Set Membership: |
|

LOC100281316
Genomic Map