Assay Details
| Gene Symbol: LOC100283359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156260.1 | NP_001149732.1 | 546 | 78 | ||
| GenBank mRNA | EU964470.1 | - | 546 | 78 | ||
| EU972594.1 | - | 94 | 78 | |||
More Information
Set Membership: |
|

LOC100283359
Genomic Map