Assay Details
| Gene Symbol: BT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130130.1 | NP_001123602.1 | 247 | 76 | ||
| GenBank mRNA | EU590667.1 | - | 247 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BT2A| Gene Symbol: BT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130130.1 | NP_001123602.1 | 247 | 76 | ||
| GenBank mRNA | EU590667.1 | - | 247 | 76 | ||
Set Membership: |
|