Assay Details
| Gene Symbol: rpl29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111474.1 | NP_001104944.1 | 648 | 169 | ||
| GenBank mRNA | AF147725.1 | - | 648 | 169 | ||
| AY107642.1 | - | 668 | 169 | |||
More Information
Set Membership: |
|

rpl29
Genomic Map