Assay Details
| Gene Symbol: tif-4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111926.1 | NP_001105396.1 | 1552 | 75 | ||
| GenBank mRNA | U73459.1 | - | 1552 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tif-4A3| Gene Symbol: tif-4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111926.1 | NP_001105396.1 | 1552 | 75 | ||
| GenBank mRNA | U73459.1 | - | 1552 | 75 | ||
Set Membership: |
|