Assay Details
| Gene Symbol: trh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112341.1 | NP_001105811.1 | 451 | 97 | ||
| GenBank mRNA | AJ890021.1 | - | 451 | 97 | ||
| EU958703.1 | - | 522 | 97 | |||
More Information
Set Membership: |
|

trh2
Genomic Map