Assay Details
| Gene Symbol: tip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111561.1 | NP_001105031.1 | 951 | 101 | ||
| GenBank mRNA | AF326502.1 | - | 951 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tip2b| Gene Symbol: tip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111561.1 | NP_001105031.1 | 951 | 101 | ||
| GenBank mRNA | AF326502.1 | - | 951 | 101 | ||
Set Membership: |
|