Assay Details
| Gene Symbol: PER70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112570.1 | NP_001106040.1 | 249 | 66 | ||
| GenBank mRNA | EF059717.1 | - | 249 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PER70| Gene Symbol: PER70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112570.1 | NP_001106040.1 | 249 | 66 | ||
| GenBank mRNA | EF059717.1 | - | 249 | 66 | ||
Set Membership: |
|