Assay Details
Gene Symbol: ras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112608.1 | NP_001106078.1 | 748 | 108 | ||
GenBank mRNA | U22432.1 | - | 748 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
