Assay Details
| Gene Symbol: fad7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111833.1 | NP_001105303.1 | 1251 | 84 | ||
| GenBank mRNA | AY107103.1 | - | 1002 | 84 | ||
| D63952.1 | - | 710 | 84 | |||
More Information
Set Membership: |
|

fad7
Genomic Map