Assay Details
| Gene Symbol: ald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111866.1 | NP_001105336.1 | 753 | 83 | ||
| GenBank mRNA | BT016836.1 | - | 828 | 83 | ||
| M16220.1 | - | 753 | 83 | |||
More Information
Set Membership: |
|

ald1
Genomic Map