Assay Details
| Gene Symbol: ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112019.1 | NP_001105489.1 | 1055 | 65 | ||
| GenBank mRNA | EU432406.1 | - | 1030 | 65 | ||
| EU432407.1 | - | 1030 | 65 | |||
| EU432408.1 | - | 1030 | 68 | |||
| EU432409.1 | - | 1030 | 68 | |||
| EU432410.1 | - | 1030 | 68 | |||
| Y13905.1 | - | 1083 | 65 | |||
More Information
Set Membership: |
|

ap1
Genomic Map