Assay Details
| Gene Symbol: mn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112126.1 | NP_001105596.1 | 151 | 73 | ||
| GenBank mRNA | AF050631.1 | - | 151 | 73 | ||
| AF165179.1 | - | 139 | 73 | |||
| AF165180.1 | - | 139 | 73 | |||
| AF165181.1 | - | 73 | 73 | |||
| AY110035.1 | - | 153 | 73 | |||
More Information
Set Membership: |
|

mn1
Genomic Map