Assay Details
| Gene Symbol: LOC100193107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138269.1 | NP_001131741.1 | 350 | 78 | ||
| GenBank mRNA | BT035279.1 | - | 225 | 78 | ||
| EU959945.1 | - | 350 | 78 | |||
More Information
Set Membership: |
|

LOC100193107
Genomic Map