Assay Details
| Gene Symbol: Atg4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144016.1 | NP_001137488.1 | 251 | 101 | ||
| GenBank mRNA | BT054942.1 | - | 243 | 101 | ||
| EU960026.1 | - | 251 | 101 | |||
| FJ444998.1 | - | 61 | 101 | |||
| FJ444999.1 | - | 61 | 101 | |||
More Information
Set Membership: |
|

Atg4a
Genomic Map