Assay Details
| Gene Symbol: LOC100282234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155146.1 | NP_001148618.1 | 1346 | 103 | ||
| GenBank mRNA | BT063406.1 | - | 1375 | 103 | ||
| EU960140.1 | - | 1346 | 103 | |||
More Information
Set Membership: |
|

LOC100282234
Genomic Map