Assay Details
| Gene Symbol: LOC100282855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155761.1 | NP_001149233.1 | 1051 | 102 | ||
| GenBank mRNA | BT068557.1 | - | 868 | 102 | ||
| EU962539.1 | - | 1051 | 102 | |||
| EU965704.1 | - | 1046 | 102 | |||
| EU966656.1 | - | 920 | 102 | |||
More Information
Set Membership: |
|

LOC100282855
Genomic Map