Assay Details
| Gene Symbol: LOC100281272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154191.1 | NP_001147663.1 | 254 | 120 | ||
| GenBank mRNA | EU956175.1 | - | 254 | 120 | ||
More Information
Set Membership: |
|

LOC100281272
Genomic Map