Assay Details
| Gene Symbol: LOC100284600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157495.1 | NP_001150967.1 | 951 | 65 | ||
| GenBank mRNA | EU968978.1 | - | 951 | 65 | ||
More Information
Set Membership: |
|

LOC100284600
Genomic Map