Assay Details
| Gene Symbol: CNR09 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154826.1 | NP_001148298.1 | 255 | 96 | ||
| GenBank mRNA | EU958331.1 | - | 255 | 96 | ||
| HM008661.1 | - | 108 | 96 | |||
More Information
Set Membership: |
|

CNR09
Genomic Map