Assay Details
| Gene Symbol: LOC100285554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158445.1 | NP_001151917.1 | 1251 | 109 | ||
| GenBank mRNA | BT042762.1 | - | 1212 | 107 | ||
| EU968417.1 | - | 1192 | 107 | |||
| EU972858.1 | - | 1251 | 109 | |||
More Information
Set Membership: |
|

LOC100285554
Genomic Map