Assay Details
| Gene Symbol: LOC100286213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159101.1 | NP_001152573.1 | 446 | 95 | ||
| GenBank mRNA | BT037891.1 | - | 391 | 95 | ||
| EU976172.1 | - | 446 | 95 | |||
More Information
Set Membership: |
|

LOC100286213
Genomic Map