Assay Details
| Gene Symbol: AC213884.3_FG001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155432.1 | NP_001148904.1 | 845 | 95 | ||
| GenBank mRNA | EU961269.1 | - | 845 | 95 | ||
More Information
Set Membership: |
|

AC213884.3_FG001
Genomic Map