Assay Details
| Gene Symbol: LOC100280921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153841.1 | NP_001147313.1 | 1846 | 93 | ||
| GenBank mRNA | EU954612.1 | - | 1846 | 93 | ||
More Information
Set Membership: |
|

LOC100280921
Genomic Map