Assay Details
| Gene Symbol: LOC100282378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155289.1 | NP_001148761.1 | 944 | 103 | ||
| GenBank mRNA | BT069920.1 | - | 929 | 103 | ||
| EU960693.1 | - | 944 | 103 | |||
More Information
Set Membership: |
|

LOC100282378
Genomic Map