Assay Details
| Gene Symbol: LOC100282213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155125.1 | NP_001148597.1 | 944 | 95 | ||
| GenBank mRNA | BT070104.1 | - | 944 | 95 | ||
| EU960051.1 | - | 934 | 95 | |||
More Information
Set Membership: |
|

LOC100282213
Genomic Map