Assay Details
| Gene Symbol: LOC100283319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156221.1 | NP_001149693.1 | 650 | 79 | ||
| GenBank mRNA | EU964285.1 | - | 650 | 79 | ||
More Information
Set Membership: |
|

LOC100283319
Genomic Map