Assay Details
| Gene Symbol: LOC100272988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147439.1 | NP_001140911.1 | 1948 | 96 | ||
| GenBank mRNA | BT039937.1 | - | 1948 | 96 | ||
More Information
Set Membership: |
|

LOC100272988
Genomic Map