Excursion-Trac Thermometers
To ensure accuracy, all Excursion-Trac Thermometers are supplied with an individually serial-numbered Traceable Certificate provided from an ISO/IEC 17025 calibration laboratory accredited by A2LA. The indicates traceability to standards provided by NIST.
- Rolling memory structure, maintains most recent 525,600 temperature observations
- Stores up to 10 unique alarm events
- Memory can be cleared on unit after downloading data to USB stick
- Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
- Status indicators - Low battery, memory full, USB data transfer and active alarm state
- Supplied: stand, Velcro™, magnetic strips, wall mount, Traceable Certificate, batteries
All Excursion-Trac Thermometers meet current CDC requirements for vaccine storage and monitoring
Excursion-Trac Datalogging Refrigerator-and-Freezer Thermometers
Excursion-Trac Datalogging Refrigerator-and-Freezer Thermometers
Traceable Memory-Loc Datalogging Thermometers
Traceable Memory-Loc Datalogging Refrigerator/Freezer Thermometers can be used in 21 CFR 11 environments.
- Use in 21 CFR 11 environments, raw data is locked and may not be cleared/changed on base unit
- Data-logging thermometer with fixed one minute logging interval
- Stores up to 10 unique alarm events
- Capable of storing over 1 million temperature observations
- Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
- Status indicators - Low battery, memory full, USB data transfer and active alarm state
- Supplied: stand, Velcro™, magnetic strips, wall mount, Traceable Certificate, batteries
All Memory-Loc Thermometers meet current CDC requirements for vaccine storage and monitoring
Memory-Loc Datalogging Refrigerator-and-Freezer Thermometers
Memory-Loc Datalogging Refrigerator-and-Freezer Thermometers
Other Refrigerator and Freezer Thermometers
Other Refrigerator and Freezer Thermometers
General Purpose Thermometers
General Purpose Thermometers
Stainless-Steel Thermometers
Stainless-Steel Thermometers
Temperature Dataloggers and Accessories
The new Fisher Scientific™ Traceable™ Logger-Trac™ Temperature Datalogger is the ultimate monitoring partner. It displays, records and reports data every step of the way. Special dataloggers and accessories are also available for measuring Relative Humidity (RH) and Liquid Nitrogen (LN2).
The Fisher Scientific Traceable Logger-Trac Temperature Dataloggers are perfect for monitoring your materials during storage, handling and transportation.
For measuring the temperature of cryo-material in transit, try the LN2 Temperature Datalogger. Or choose the mobile Relative Humidity and Temperature Datalogger. Optional accessories include waterproof sleeves, a cable-and-cradle combination, a USB cable and MaxiThermal software.
Fisher Scientific Traceable Logger-Trac Brochure
Fisher Scientific Traceable Logger-Trac Thermometer is ideal for monitoring material during storage, handling, transportation.