Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 1-EBIO
GSA_VASDP

Catalog No. 104150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-415-0 50 mg
10-411-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-415-0 Supplier R&D Systems™ Supplier No. 1041/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Activator of epithelial KCa channels

Activator of epithelial KCa channels, stimulates a large and sustained trans-epithelial Cl- secretory response across T84 monolayers. Induces hyperpolarization to the same magnitude as ACh in aortic value endothelial cells. Promotes embryonic stem cell (ESC) differentiation into cardiomyocytes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 10045-45-1
Molecular Formula C9H10N2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 162.19
Purity >99%
Molecular Weight (g/mol) 162.19
Chemical Name or Material 1-EBIO
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.