CAS RN 1009816-48-1
IUPAC Name:
2-(ethylamino)-5-(hydroxymethyl)-3aH,5H,6H,7H,7aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
Synonyms:
2-(ethylamino)-5-(hydroxymethyl)-3aH,5H,6H,7H,7aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
Molecular Weight (g/mol):
248.3
Molecular Formula:
C9H16N2O4S
InChi Key:
PPAIMZHKIXDJRN-UHFFFAOYNA-N
SMILES:
CCNC1=NC2C(OC(CO)C(O)C2O)S1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ O-GlcNAcase Inhibitor, TMG
A cell-permeable, blood-brain barrier-permeant, orally active aminothiazoline that effectively increases O -GlcNAc level of cellular proteins both in cultures (EC50 = 30 nM, ECmax = 200nM in PC3) in vitro and in rats
2
R&D Systems™ Thiamet G
CAS: 1009816-48-1 Molecular Formula: C9H16N2O4S