Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Thiamet G
GREENER_CHOICESDP

Catalog No. 439010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-901-0 Supplier R&D Systems™ Supplier No. 4390/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent O-GlcNAcase inhibitor

Potent, selective inhibitor of O-GlcNAcase (Ki = 21 nM for human O-GlcNAcase). Decreases the phosphorylation of tau protein in vivo. Promotes autophagy independently of mTOR pathway and reduces toxic protein species in mouse tauopathy model. Orally bioavailable and blood brain barrier permeable.

Chemical Identifiers

CAS 1009816-48-1
Molecular Formula C9H16N2O4S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 248.3
Purity >98%
Molecular Weight (g/mol) 248.297
Chemical Name or Material Thiamet G
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.