accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-38-2

ClNClOCl

CAS RN 101-38-2

IUPAC Name: 2,6-dichloro-4-(chloroimino)cyclohexa-2,5-dien-1-one
Synonyms: gibbs reagent
Molecular Weight (g/mol): 210.44
Molecular Formula: C6H2Cl3NO
InChi Key: YHUMTHWQGWPJOQ-UHFFFAOYSA-N
SMILES: ClN=C1C=C(Cl)C(=O)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results