CAS RN 10124-41-1
CAS RN 10124-41-1
IUPAC Name:
calcium 1-oxo-1λ⁶-disulfene-1,1-bis(olate)
Synonyms:
calcium Hypo
Molecular Weight (g/mol):
152.2
Molecular Formula:
CaO3S2
InChi Key:
FAYYUXPSKDFLEC-UHFFFAOYSA-L
SMILES:
[Ca++].[O-]S([O-])(=O)=S
1
–
1
of
1
results
1
Thermo Scientific Chemicals Calcium thiosulfate, pure, 30-50% solution in water
CAS: 10124-41-1 | CaO3S2 | 152.21 g/mol