accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1019-71-2

IUPAC Name: chlorobis(4-methylphenyl)phosphane
Synonyms: chlorobis(4-methylphenyl)phosphane
Molecular Weight (g/mol): 248.69
Molecular Formula: C14H14ClP
InChi Key: BJBXRRHIBSXGLF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)P(Cl)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)