accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1021868-83-6

IUPAC Name: [dibromo({[({5-[6-(diethylamino)-9H-purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy)(hydroxy)phosphoryl]oxy}(hydroxy)phosphoryl)methyl]phosphonic acid trisodium
Synonyms: dibromo[({5-[6-(diethylamino)purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy(hydroxy)phosphoryl)oxy(hydroxy)phosphoryl]methylphosphonic acid trisodium
Molecular Weight (g/mol): 788.08
Molecular Formula: C15H24Br2N5Na3O12P3
InChi Key: BBYLOOHIKBMBIW-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].CCN(CC)C1=C2N=CN(C3OC(COP(O)(=O)OP(O)(=O)C(Br)(Br)P(O)(O)=O)C(O)C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results