Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ARL 67156 trisodium salt
GREENER_CHOICESDP

Catalog No. 128310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-831-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-831-0 Supplier Tocris Bioscience™ Supplier No. 1283/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NTPDase inhibitor

ARL 67156 trisodium salt is a selective NTPDase inhibitor (pIC50 = 4.62 and 5.1 in human blood and rat vas deferens respectively). ARL 67156 prevents vascular calcification and extends longevity in Hutchinson-Gilford progeria syndrome mice. ARL 67156 increases neurogenic contract response to nerve stimulation. ARL 67156 enhances parasympathetic neurotransmission in guinea-pig urinary bladder.

Chemical Identifiers

CAS 1021868-83-6
Molecular Formula C15H24Br2N5Na3O12P3

Specifications

Quantity 10 mg
Formula Weight 785.06
Purity >98%
Molecular Weight (g/mol) 788.079
Chemical Name or Material ARL 67156 trisodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.