accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(R)H3CH3C(R)H3COHN(S)CH3ONH3C(R)BrHNOHN(R)OOOH(E)

CAS RN 102396-24-7

IUPAC Name: (4R,7R,10S,13R,15E,17R,19S)-7-[(2-bromo-1H-indol-3-yl)methyl]-4-(4-hydroxyphenyl)-8,10,13,15,17,19-hexamethyl-1-oxa-5,8,11-triazacyclononadec-15-ene-2,6,9,12-tetrone
Synonyms: (4R,7R,10S,13R,15E,17R,19S)-7-[(2-bromo-1H-indol-3-yl)methyl]-4-(4-hydroxyphenyl)-8,10,13,15,17,19-hexamethyl-1-oxa-5,8,11-triazacyclononadec-15-ene-2,6,9,12-tetrone
Molecular Weight (g/mol): 709.68
Molecular Formula: C36H45BrN4O6
InChi Key: GQWYWHOHRVVHAP-HZIRKPFGSA-N
SMILES: C[C@H]1C[C@@H](C)\C=C(C)\C[C@@H](C)C(=O)N[C@@H](C)C(=O)N(C)[C@H](CC2=C(Br)NC3=CC=CC=C23)C(=O)N[C@H](CC(=O)O1)C1=CC=C(O)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)