accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNH2NNH2OH3COCH3

CAS RN 1026582-88-6

IUPAC Name: 5-{[4,5-dimethoxy-2-(propan-2-yl)phenyl]methyl}pyrimidine-2,4-diamine
Synonyms: 5-[(2-isopropyl-4,5-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol): 302.38
Molecular Formula: C16H22N4O2
InChi Key: PYNPWUIBJMVRIG-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C(C)C)C(CC2=CN=C(N)N=C2N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)