Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ RO-3
GREENER_CHOICESDP

Catalog No. 305210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-5210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-5210 Supplier R&D Systems™ Supplier No. 3052/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective P2X3 and P2X2/3 antagonist

Selective homomeric P2X3 and heteromeric P2X2/3 receptor antagonist (pIC50 values are 7.0 and 5.9 respectively) that exhibits no activity at P2X1, P2X2, P2X4, P2X5 and P2X7 receptors (IC50 > 10 μM). Attenuates nociceptive sensitivity in animal models of pain. Orally active and brain penetrant.

Chemical Identifiers

CAS 1026582-88-6
Molecular Formula C16H22N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 302.37
Purity >98%
Molecular Weight (g/mol) 302.378
Chemical Name or Material RO-3
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.