CAS RN 103-90-2
4-Acetamidophenol, 98%
CAS: 103-90-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD00002328 InChI Key: RZVAJINKPMORJF-UHFFFAOYSA-N Synonym: acetaminophen,4-acetamidophenol,paracetamol,apap,n-4-hydroxyphenyl acetamide,tylenol,acetaminofen,panadol,datril,p-acetamidophenol PubChem CID: 1983 ChEBI: CHEBI:46195 IUPAC Name: N-(4-hydroxyphenyl)acetamide SMILES: CC(=O)NC1=CC=C(O)C=C1
4-Acetamidophenol, 98%
CAS: 103-90-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD00002328 InChI Key: RZVAJINKPMORJF-UHFFFAOYSA-N Synonym: acetaminophen,4-acetamidophenol,paracetamol,apap,n-4-hydroxyphenyl acetamide,tylenol,acetaminofen,panadol,datril,p-acetamidophenol PubChem CID: 1983 ChEBI: CHEBI:46195 IUPAC Name: N-(4-hydroxyphenyl)acetamide SMILES: CC(=O)NC1=CC=C(O)C=C1
CERILLIANT™ Acetaminophen Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
Suitable for use as starting material for calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to P450 studies and other pharmaceutical research.
Acetaminophen, Powder, USP, 98-102%, Spectrum™ Chemical
CAS: 103-90-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD00002328 InChI Key: RZVAJINKPMORJF-UHFFFAOYSA-N IUPAC Name: N-(4-hydroxyphenyl)acetamide SMILES: CC(=O)NC1=CC=C(O)C=C1
Acetaminophen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Acetaminophen is a drug, which possesses pharmacological properties like antipyretic and analgesic property.
4'-Hydroxyacetanilide 98.0+%, TCI America™
CAS: 103-90-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD00002328 InChI Key: RZVAJINKPMORJF-UHFFFAOYSA-N Synonym: acetaminophen,4-acetamidophenol,paracetamol,apap,n-4-hydroxyphenyl acetamide,tylenol,acetaminofen,panadol,datril,p-acetamidophenol PubChem CID: 1983 ChEBI: CHEBI:46195 IUPAC Name: N-(4-hydroxyphenyl)acetamide SMILES: CC(=O)NC1=CC=C(O)C=C1
R&D Systems™ Acetaminophen
CAS: 103-90-2 Molecular Formula: C8H9NO2
Medisca Acetaminophen, USP
Acetaminophen USP is a white, odorless, crystalline powder. Alternative name(s): paracetamol