accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOHNFFFFNHNHOHN(E/Z)

CAS RN 1033769-28-6

IUPAC Name: 1-[2-fluoro-5-(trifluoromethyl)phenyl]-3-[4-methyl-3-({2-oxo-3-[(1H-pyrrol-2-yl)methylidene]-2,3-dihydro-1H-indol-6-yl}amino)phenyl]urea
Synonyms: 1-[2-fluoro-5-(trifluoromethyl)phenyl]-3-(4-methyl-3-{[2-oxo-3-(1H-pyrrol-2-ylmethylidene)-1H-indol-6-yl]amino}phenyl)urea
Molecular Weight (g/mol): 535.5
Molecular Formula: C28H21F4N5O2
InChi Key: YYDUWLSETXNJJT-UHFFFAOYSA-N
SMILES: CC1=C(NC2=CC3=C(C=C2)C(=CC2=CC=CN2)C(=O)N3)C=C(NC(=O)NC2=C(F)C=CC(=C2)C(F)(F)F)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)