Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GNF 5837
GSA_VAGREENER_CHOICESDP

Catalog No. 455950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-595-0 50 mg
45-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-595-0 Supplier R&D Systems™ Supplier No. 4559/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent Trk inhibitor; inhibits TrkA, TrkB and TrkC

Potent pan-Trk inhibitor. Displays antiproliferative effects in cellular Ba/F3 assays (IC50 values are 7, 9 and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively). Exhibits selectivity for Trk receptors over a range of kinases, with some activity at PDGFR and c-Kit (IC50 values are 0.87 and 0.91 μM respectively). Orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1033769-28-6
Molecular Formula C28H21F4N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 535.49
Purity >98%
Molecular Weight (g/mol) 535.50
Chemical Name or Material GNF 5837
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.