Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Caffeic acid phenethyl ester
GREENER_CHOICESDP

Catalog No. 274350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-435-0 50 mg
27-431-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-435-0 Supplier R&D Systems™ Supplier No. 2743/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Specific inhibitor of NF-κB activation

Antioxidant, antimitogenic, anticarcinogenic, anti-inflammatory and antiviral. Specifically inhibits NF-κB activation and inhibits the growth of transformed cells. Also suppresses lipid peroxidation and inhibits ornithine decarboxylase, protein tyrosine kinase and lipoxygenase activities.

Chemical Identifiers

CAS 104594-70-9
Molecular Formula C17H16O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 284.31
Purity 98%
Molecular Weight (g/mol) 284.31
Chemical Name or Material Caffeic acid phenethyl ester
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.