accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNClNNNO

CAS RN 104615-18-1

IUPAC Name: 9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine
Synonyms: 9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine
Molecular Weight (g/mol): 285.69
Molecular Formula: C13H8ClN5O
InChi Key: MSJODEOZODDVGW-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(Cl)C=C2C2=NC(=NN12)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)